missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Magnetic Retrievers for Stirrer Bars

Retrieve stirrer bars from sensitive liquids with these magnetic retrievers. Azlon™ Magnetic Retrievers for Stirrer Bars feature magnets permanently embedded in a PTFE rod.

Brand:  Azlon™ SWN564

28.28 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10514355

  • £44.90 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Retrieve stirrer bars from sensitive liquids with these magnetic retrievers. AzlonTM Magnetic Retrievers for Stirrer Bars feature magnets permanently embedded in a PTFE rod.

  • Ideal for retrieving magnetic stirrer bars from particularly sensitive or corrosive liquids
  • Turbo retrievers are ideal for use with turbo stirrer bars
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
98 in.
10 mm
Magnet
Retrievers Stirrer Bar
98 Lin.
PTFE
250 mm
1/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.