missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azlon™ Pivot Ring Magnetic Stirrer Bar
Constructed of magnetic PTFE. Azlon™ Pivot Ring Stirrer Bar includes a central pivot ring that ensures efficient spinning even on curved or uneven bases.
Brand: Azlon SWN511
9.13 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Description
Constructed of magnetic PTFE. Azlon™ Pivot Ring Stirrer Bar includes a central pivot ring that ensures efficient spinning even on curved or uneven bases.
- Constructed of magnetic PTFE
- Central pivot ring ensures efficient spinning even on curved or uneven bases
Specifications
Autoclavable | |
1.18 in. | |
6 mm | |
Magnet | |
Pivot ring |
1.18 Lin. | |
PTFE | |
30 mm | |
2/Pk. | |
Stir Bar |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction