Learn More
Azlon™ Pivot Ring Magnetic Stirrer Bar
Constructed of magnetic PTFE. Azlon™ Pivot Ring Stirrer Bar includes a central pivot ring that ensures efficient spinning even on curved or uneven bases.
£8.15 - £27.15
Specifications
Autoclavable | Yes |
---|---|
Coating | PTFE |
Material | Magnet |
Quantity | 2/Pk. |
Shape | Pivot ring |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
10648195
|
Azlon
SWN502 |
12 mm | 4.5 mm |
£10.30
Pack of 2 |
In Stock
Log in to see stock. |
||||
10298094
|
Azlon
SWN501 |
15 mm | 4.5 mm |
£11.51
Pack of 2 |
In Stock
Log in to see stock. |
||||
10585105
|
Azlon
SWN503 |
20 mm | 6 mm |
£11.76
Pack of 2 |
In Stock
Log in to see stock. |
||||
10135374
|
Azlon
SWN504 |
25 mm | 6 mm |
£11.00
Pack of 2 |
In Stock
Log in to see stock. |
||||
10729175
|
Azlon
SWN511 |
30 mm | 6 mm |
£13.83
Pack of 2 |
In Stock
Log in to see stock. |
||||
10636855
|
Azlon
SWN506 |
45 mm | 8 mm |
£17.29
Pack of 2 |
In Stock
Log in to see stock. |
||||
10431235
|
Azlon
SWN507 |
40 mm | 8 mm |
£15.38
Pack of 2 |
In Stock
Log in to see stock. |
||||
10145374
|
Azlon
SWN505 |
35 mm | 6 mm |
£16.05
Pack of 2 |
In Stock
Log in to see stock. |
||||
10788995
|
Azlon
SWN509 |
50 mm | 8 mm |
£18.94
Pack of 2 |
In Stock
Log in to see stock. |
||||
10431425
|
Azlon
SWN508 |
60 mm |
£24.30
Pack of 2 |
In Stock
Log in to see stock. |
|||||
10564345
|
Azlon
SWN510 |
70 mm | 9.5 mm |
£27.15
Pack of 2 |
In Stock
Log in to see stock. |
||||
10738795
|
Azlon
SWN500 |
8 mm | 3 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Constructed of magnetic PTFE. Azlon™ Pivot Ring Stirrer Bar includes a central pivot ring that ensures efficient spinning even on curved or uneven bases.
- Constructed of magnetic PTFE
- Central pivot ring ensures efficient spinning even on curved or uneven bases
Specifications
Yes | |
Magnet | |
Pivot ring |
PTFE | |
2/Pk. | |
Stir Bar |