Learn More
Azlon™ Polyamide Coated Steel Bottle Carrier
Ideal for safely carrying up to six bottles or wash bottles
Brand:  Azlon BGS040
 View more versions of this product
Product Code. 12030832
  • £179.00 / Each
Estimated Shipment: 03-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from polyamide coated steel
  • Offers better heat and chemical resistance than other coatings
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Bottle Carrier
Polyamide-coated Steel
1
60 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Azlon™ Polyamide Coated Steel Bottle Carrier > Max bottle diameter: 60mm