missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azlon™ Polypropylene Measuring Scoop
Scoop chemical reagents or food items safely. The Azlon™ polypropylene measuring scoop comes with flat top and bottom.
Brand: Azlon SWL012
12.20 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Scoop chemical reagents or food items safely. The Azlon™ polypropylene measuring scoop comes with flat top and bottom.
- Made from food safe polypropylene
- Top is flat for levelling off powders
- Base is flat enabling scoops to stand horizontally
- The capacity is clearly molded on each size
Specifications
Measuring Scoop | |
Handle | |
137 mm |
Polypropylene | |
25 mL |