missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Polypropylene Measuring Scoop

Scoop chemical reagents or food items safely. The Azlon™ polypropylene measuring scoop comes with flat top and bottom.

Brand:  Azlon SWL140

25.04 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11349633

  • £32.65 / Each
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Scoop chemical reagents or food items safely. The Azlon™ polypropylene measuring scoop comes with flat top and bottom.

  • Made from food safe polypropylene
  • Top is flat for levelling off powders
  • Base is flat enabling scoops to stand horizontally
  • The capacity is clearly molded on each size
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Measuring Scoop
Handle
385 mm
Polypropylene
1000 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers