missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azlon™ Tapered Glass Joint Sleeve
Forms a liquid and gas tight seal.
Brand: Azlon SWN810
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
- For grease free and high vacuum seals using Quickfit™ ground glass joints
- Strong construction with knurled shoulder for ease of handling and repeated use
- Self lubricating and non-contaminating
- Working temperature range -100°C to +260°C
- Working vacuum ≤10-3 atmospheres
- Highly resistant to chemical attack
Spécification
PTFE | |
Tapered sleeves | |
–100 to 250°C | |
29/32 |