missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azpack™ Mortar and Pestle with Wooden Handle Pestle
Releases oils that can't be extracted using modern appliances. Azpack™ Mortar and Pestle with Wooden Handle will grind and crush the toughest materials without cracking the strong and dense porcelain.
Brand: Azpack™ 12393338
This item is not returnable.
View return policy
Description
- Mortar constructed of vitrified electical porcelain
- Pestle includes a Beech wood handle
- Unglazed grinding surface
- Integral pouring lip reduces waste and spillage
Specifications
199 mL | |
95mm | |
Mortar and Pestle | |
74mm (Mortar) |
Unglazed | |
113mm (118mm to lip) | |
Mortar: Vitrified electrical porcelain, Pestle: Beech | |
170mm (Pestle) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction