Learn More
Thermo Scientific Chemicals Azur eosin methylene-blue, pure, solution according to Giemsa
Brand: Thermo Scientific Chemicals 295591000
15.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Azur eosin methylene-blue | |
MFCD00081642 | |
Pure | |
Blue | |
Solution |
51811-82-6, 67-56-1 | |
523 to 526nm (c=0.1, MeOH) (eosine), 646 to 651nm (c=0.1, MeOH) (methylene blue) | |
Glass bottle | |
100 mL |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- Azur eosin methylene-blue solution according to Giemsa
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H311-Toxic in contact with skin.
- H301-Toxic if swallowed.
- H370-Causes damage to organs.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : Methyl alcohol, Giemsa's stain
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0467139 | Certificate of Analysis | 30/01/2025 | 295591000,295595000 |
A0464345 | Certificate of Analysis | 09/07/2024 | 295591000,295595000 |
A0463876 | Certificate of Analysis | 20/06/2024 | 295591000,295595000 |
A0458753 | Certificate of Analysis | 14/12/2023 | 295591000,295595000 |
A0451970 | Certificate of Analysis | 02/05/2023 | 295591000,295595000 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only