Learn More
Gibco™ B-27™ Supplement, minus insulin
B-27™ Supplement, minus insulin
Brand: Gibco™ A1895601
81.64 GBP valid until 2025-12-31
Use promo code "24342" to get your promotional price.
Description
B-27 Supplement is suitable for use in most neuronal cell applications. For other applications, we provide alternative formulations of B-27 Supplement and Neurobasal media.
For established protocols using B-27 Supplement, access our Gibco Neurobiology Protocol Handbook.
View publications using Gibco neurobiology products at our Gibco CellCite.

Specifications
Store in freezer (-5 to -20°C). Protect from light. | |
Serum-free | |
Mammalian Cell Culture | |
Cell Culture Supplement | |
Sterile | |
10 mL | |
Serum-Free |
Neural Cells | |
50 X | |
Liquid | |
Serum-Free | |
B-27™ | |
Dry Ice | |
Endotoxin, Performance, Sterility, Mycoplasma |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2415097 | Certificate of Analysis | 29/01/2025 | A1895601,A1895602 |
2445009 | Certificate of Analysis | 29/01/2025 | A1895601,A1895602 |
2415091 | Certificate of Analysis | 29/01/2025 | A1895601,A1895602 |
2442734 | Certificate of Analysis | 29/01/2025 | A1895601,A1895602 |
2445015 | Certificate of Analysis | 29/01/2025 | A1895601,A1895602 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.