Learn More
Brady™ B-423 Polyester Thermal Transfer Labels
Printable, polyester labels for Thermal Transfer Printers.
Brand:  Brady™ 030357
177.88 GBP valid until 2024-12-20
Use promo code "21731" to get your promotional price.
 View more versions of this product
Product Code. 16677865
  • £249.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Permanent adhesive
  • For smooth surfaces
Specifications
Specifications
Autoclavable
Permanent Acrylic
Gloss
Thermal Transfer
Barcode Labeling, Circuit Board Labeling, Equipment Labeling, General Identification, Inventory Labeling
1
Rectangle
0.076 mm
50.8 mm
10.2 mm
UL, CSA and AGA Approved
White
Polyester (B-423)
Non-sterile
BBP™81, BradyPrinter i5100, i7100, PR Plus Series, and IP™ Series Printers, THT 76 mm Core Printers
10,000 Labels
-70°C to +110°C
12.7 mm
50.8 mm
10.2 mm