Learn More
Brady™ B33 Series Polyamide Coated Nylon Cloth Labels
Nylon cloth labels for BBP33/i3300 printers.
Brand:  Brady™ 361663
Additional Details : Weight : 0.44700kg
 View more versions of this product
Product Code. 30041401
  • £204.81 / Pack of 3000
In Stock
to see stock.
Description
Description
For BBP33/i3300 printers.
Specifications
Specifications
UL Recognized,CSA Accepted;Halogen Free (DIN VDE 0472/part 815) Approvals are for White material only
Permanent Acrylic
Matte
Labels
Component and Equipment Labelling, Electrical Labelling, General Identification, Laboratory Labelling, Warehouse Marking, Wire and Cable Labelling
3
Round
-40°C to +90°C
0.166 mm
9.53 mm (Dia)
9.53 mm (Dia)
9.53 mm
Nylon Cloth Labels for BBP33/i3300 Printers
White
Nylon Cloth
Non-sterile
BBP33, i3300,i5300
300 Stickers
-40°F to +194°F
0.0065 in.
9.53 mm (Dia)
9.53 mm (Dia)
0.375 in.