Learn More
CPC™ Back-to-Back Insert Adapter
MPC to MPX Back-to-Back Insert Adapter
Brand:  CPC™ MPC22X2239M
Product Code. 17874454
  • £469.49 / Pack of 25
Explore more special offers
Description
Description
  • Medical Grade
  • Non-valved
  • Seal Options: Silicone, platinum-cured, USP Class VI, ADCF
Specifications
Specifications
USP Class VI, ADCF
Polysulfone
Gamma, Autoclave
-40°F
Amber
Up to 60 psi, 4.1 bar
300°F
149°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
CPC™ Back-to-Back Insert Adapter > Polysulfone, USP Class VI, ADCF, sterlizations: Gamma Autoclave, free floating, -40 F to 300 F