missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Bacto™ Yeast Extract, technical
Suitable for use as multi-functional nutritional supplements in mammalian cell culture, microbial fermentation, and insect cell culture applications.
Brand: Gibco™ 288620
This item is not returnable.
View return policy
Description
- Concentrates of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells derived from primary grown baker's yeast
- These animal origin-free products are suitable for use as multi-functional nutritional supplements in mammalian cell culture, microbial fermentation, and insect cell culture applications
- Yeast extract provides essential water-soluble vitamins, amino acids, peptides, and carbohydrates to any medium formulation
- Bacto Yeast Extract is an important ingredient for the microbiological assay of vitamins
- B factor, a growth substance necessary for the production of rifampin in a Nocardia species, can be isolated from yeast extract
- Developed to provide products priced for the biotechnology market with acceptable clarity and growth-promoting characteristics
- Store in cool, dry conditions in well-sealed containers at 2°C to 25°C

Specifications
LUO | |
Mammalian, Insect, Microbial | |
Bacterial Media | |
Dehydrated culture media | |
500 g |
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. | |
Dehydrated | |
Bottle | |
Bacto™ | |
Animal origin-free |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ Bacto™ Yeast Extract, technical > 500 g
Spot an opportunity for improvement?Share a Content Correction