missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Yeast Extract (Granulated)

Dried yeast autolysate with a high content of amino nitrogen and water soluble B complex vitamins

Brand:  Fisher BioReagents™ BP9727-500

47.13 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 11385992

  • £108.00 / 500g

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Dried yeast autolysate with a high content of amino nitrogen and water soluble B complex vitamins

  • Water-soluble portion of autolyzed yeast containing vitamin B complex
  • Used in preparing microbiological culture media
  • CAS: 8013-01-2

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dehydrated
Microbiology Media
Vitamin B complex
500 g
Yeast Extract
Used for preparing microbiological culture media
Poly Bottle
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Yeast Extract

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisher BioReagents™ Yeast Extract (Granulated) > 500g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.