missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Yeast Extract (Granulated)

Dried yeast autolysate with a high content of amino nitrogen and water soluble B complex vitamins

£108.00 - £329.00

Specifications

Form Dehydrated
Media Type Yeast Extract
Includes Vitamin B complex
View More Specs

Products 2
Product Code Brand Quantity Packaging Type Media Type Price Quantity & Availability  
Product Code Brand Quantity Packaging Type Media Type Price Quantity & Availability  
11385992
View Documents
Fisher BioReagents™
BP9727-500
500 g Poly Bottle Yeast Extract
£108.00
500g
In Stock
to see stock.
Edge
Add to basket
 
11365992
View Documents
Fisher BioReagents™
BP9727-2
2000 g Poly Bottle Yeast Extract
£329.00
2kg
In Stock
to see stock.
Add to basket
 
Description

Description

Dried yeast autolysate with a high content of amino nitrogen and water soluble B complex vitamins

  • Water-soluble portion of autolyzed yeast containing vitamin B complex
  • Used in preparing microbiological culture media
  • CAS: 8013-01-2

Specifications

Specifications

Dehydrated
Yeast Extract
Vitamin B complex
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Yeast Extract

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisher BioReagents™ Yeast Extract (Granulated)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.