Learn More
J.T. Baker™ BAKER spe-12G Column Processor, Glass Design
Offers the flexibility of processing up to 12 SPE devices of different heights, diameters, or formats at the same time. J.T. Baker™ BAKER spe-12G Column Processor, Glass Design is a complete 12-port system that comes complete with a glass vacuum basin and sample collection rack.
Brand:  J.T. Baker™ 6998-00
Product Code. 15597644
  • £2950.00 / Each
Estimated Shipment: 15-08-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The design of the BAKER spe-12G Column Processor, Glass Design is familiar throughout the industry, and supports all devices and accessories with luer-type fittings (e.g. BAKERBOND spe, BAKERBOND Speedisk™ columns, and BAKERBOND Speedisk™ extraction disks).
The J.T. Baker™ BAKER spe-12G is a 12-port system, suitable for processing up to 12 SPE columns at the same time. The processor comes complete with:
  • glass vacuum basin
  • cover with luer fittings and gasket
  • individual flow control stopcocks
  • stainless steel needles
  • glass sample collection rack
  • height-adjustable shelves
  • vacuum gauge/controller
Specifications
Specifications
Solid Phase Extraction
SPE Vacuum Processor
1 Each
Extraction Columns and Discs with Luer-Type Fittings, Such as BAKERBOND Spe, BAKERBOND Speedisk Colu
Glass vacuum chamber and sample collection rack, polyamide lid, PTFE luer connectors/stopcocks, polyethylene/neoprene gaskets
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
J.T. Baker™ BAKER spe-12G Column Processor, Glass Design > Glass Design