missing translation for 'onlineSavingsMsg'
Learn More

Avantor™ BAKERBOND™ spe Octadecyl (C18) Disposable Extraction Columns

Define and control critical surface chemistry parameters to ensure performance consistency. Avantor BAKERBOND spe Octadecyl (C18) Disposable Extraction Columns, are designed with standard geometries to adapt to most popular automated liquid handling systems for extracting non-ionic, non-polar to moderately polar analytes.

Brand:  Avantor™ 7020-03

Code : Z3

Additional Details : CAS Number : 71889-02-6 Weight : 0.10000kg

 View more versions of this product

Product Code. 13104704

  • POA / Pack of 50
Estimated Shipment: 19-08-2024
to see stock.

This item requires a price update prior to being ordered.

Request a Price for this item here.


Description

Description

  • Constructed of ultraclean polypropylene
  • Provide recoveries when extracting analytes of weak hydrophobicity
  • Consistent bonding chemistry
  • Wide range of formats
Specifications

Specifications

17.2%
Quantity: 50/Pk.
Non-Ionic, Non-Polar to Moderately Polar Analytes
C18, Octadecyl
Yes
3 mL
500 mg
BAKER SPE-12 G Column Processor Complete (6998-00) or BAKER SPE-24 G Column Processor Complete (7208
Solid Phase Extraction Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers