Learn More
Cole-Parmer™ Balance Draft Shield
Clear acrylic prevents drafts from affecting weighings
£179.00 - £378.56
Specifications
Color Clear
View More Specs
Products 3
Product Code Brand Width (Metric) Price Quantity & Availability  
Product Code Brand Width (Metric) Price Quantity & Availability  
15380645
View Documents
Cole-Parmer™
01019-02
330 mm
£179.00
Each
12891763
View Documents
Cole-Parmer™
01019-04
381 mm
£213.00
Each
15300655
View Documents
Cole-Parmer™
01019-06
457 mm
£378.56
Each
Description
Description
Clear acrylic prevents drafts from affecting weighings
  • No assembly required
  • Mouse hole in back panel for power cords
  • Raised edges on top of enclosure allow secure placement of frequently used weighing accessories
  • Minimizes air movement around your balance for faster, more stable readings
Specifications
Specifications
Clear
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Cole-Parmer™ Balance Draft Shield