Learn More
Cole-Parmer™ Balance Draft Shield
Clear acrylic prevents drafts from affecting weighings
Brand:  Cole-Parmer™ 01019-04
 View more versions of this product
Product Code. 12891763
  • £213.00 / Each
Explore more special offers
Description
Description
Clear acrylic prevents drafts from affecting weighings
  • No assembly required
  • Mouse hole in back panel for power cords
  • Raised edges on top of enclosure allow secure placement of frequently used weighing accessories
  • Minimizes air movement around your balance for faster, more stable readings
Specifications
Specifications
Clear
381 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ Balance Draft Shield > Dimensions: 425D x 381W x 394mmH, Size: Medium