Learn More
Kinesis™ Barbed Adapter
Thread to barbed, flat bottom
Brand:  Kinesis™ 008NB20-KD5L
Encompass_Preferred
 View more versions of this product
Product Code. 13106844
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Color: Natural
  • PEEK
  • Flat bottom
  • Specifications
    Specifications
    2 mm
    PEEK
    1/4-28M