missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium chloride dihydrate, Puriss. p.a. plus, ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
5 kg
4 x 5 kg
50 kg
6 x 1 kg
Description
- Exceeds standard ACS specifications with additional trace metals analysis
- Products with a guarantee certificate and/or suitable for the stated analytical application.

Specifications
Specifications
Quantity | 100 g |
UN Number | 1564 |
Formula Weight | 244.26g/mol |
Percent Purity | ≥99% |
Grade | ACS/puriss. p.a. plus |
Packaging | Plastic Bottle |
Chemical Name or Material | Barium Chloride Dihydrate, Reag. Ph. Eur. reag. ISO |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 6
Safety and Handling

Product Identifier
- Barium chloride dihydrate
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- H301-Toxic if swallowed.
- H332-Harmful if inhaled.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Barium chloride dihydrate, Puriss. p.a. plus, ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%, Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction