missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium chloride dihydrate, Puriss. p.a. plus, ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%, Honeywell Fluka™
Puriss. p.a. plus, ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99%
£50.11 - £1020.25
Chemical Identifiers
CAS | 10326-27-9 |
---|---|
Molecular Formula | BaCl2H4O2 |
Molecular Weight (g/mol) | 244.26 |
MDL Number | MFCD00149154 |
InChI Key | PWHCIQQGOQTFAE-UHFFFAOYSA-L |
Synonym | barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d |
PubChem CID | 5284346 |
ChEBI | CHEBI:86153 |
IUPAC Name | barium(2+);dichloride;dihydrate |
SMILES | O.O.[Cl-].[Cl-].[Ba++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15633250
|
Honeywell-Fluka
31125-100G |
100 g |
£62.50
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15673250
|
Honeywell-Fluka
31125-500G |
500 g |
£50.11
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15643250
|
Honeywell-Fluka
31125-1KG |
1 kg |
£77.29
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15653250
|
Honeywell-Fluka
31125-2.5KG |
2.5 kg | |||||||
15693250
|
Honeywell-Fluka
31125-5KG |
5 kg |
£257.76
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15603260
|
Honeywell-Fluka
31125-6X1KG |
6 x 1 kg |
£378.11
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15663250
|
Honeywell-Fluka
31125-4X5KG |
4 x 5 kg |
£995.24
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15683250
|
Honeywell-Fluka
31125-50KG |
50 kg | |||||||
Description
- Exceeds standard ACS specifications with additional trace metals analysis
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Chemical Identifiers
10326-27-9 | |
244.26 | |
PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
5284346 | |
barium(2+);dichloride;dihydrate |
BaCl2H4O2 | |
MFCD00149154 | |
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
CHEBI:86153 | |
O.O.[Cl-].[Cl-].[Ba++] |
Specifications
10326-27-9 | |
MFCD00149154 | |
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
O.O.[Cl-].[Cl-].[Ba++] | |
244.26 | |
CHEBI:86153 | |
≥99% | |
Plastic Bottle |
BaCl2H4O2 | |
1564 | |
PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
barium(2+);dichloride;dihydrate | |
5284346 | |
244.26g/mol | |
ACS/puriss. p.a. plus | |
Barium Chloride Dihydrate, Reag. Ph. Eur. reag. ISO |
Safety and Handling
P261
H301-H332
missing translation for 'einecsNumber' : 233-788-1
missing translation for 'rtecsNumber' : CQ8751000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Barium chloride dihydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
Hazard Statement
- H301-Toxic if swallowed.
- H332-Harmful if inhaled.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.