Learn More
Barium hydroxide, anhydrous, 94-98%, Thermo Scientific Chemicals
£28.74 - £108.54
Chemical Identifiers
CAS | 17194-00-2 |
---|---|
Molecular Formula | BaH2O2 |
Molecular Weight (g/mol) | 171.341 |
MDL Number | MFCD00003443 |
InChI Key | RQPZNWPYLFFXCP-UHFFFAOYSA-L |
Synonym | barium hydroxide, barium 2+ dihydroxide, bariumhydroxid, aetzbaryt, barium ii hydroxide, barium 2+ hydroxide, acmc-20alr7, barium hydroxide,anhydrous, barium hydroxide anhydrous |
PubChem CID | 6093286 |
ChEBI | CHEBI:32592 |
IUPAC Name | barium(2+);dihydroxide |
SMILES | [OH-].[OH-].[Ba+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11396588
|
Thermo Scientific Alfa Aesar
012195.18 |
50 g |
£28.74
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11306598
|
Thermo Scientific Alfa Aesar
012195.30 |
250 g |
£40.20
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11316598
|
Thermo Scientific Alfa Aesar
012195.A1 |
1 kg |
£108.54
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cobalt(II,III) oxide is used in enamels, pigments, porcelain colorants, semiconductors, ceramics, catalysis, grinding wheels and other fields as an important inorganic material. It is a useful oxidant in chemical industry and as catalyst for organic synthesis. It is widely used as catalyst carrier and as electrode active material. It is also used in senior goggles, other filter materials, in temperature and gas sensors, in electronic ceramics, in lithium ion battery electrode materials, magnetic materials, electrochromic devices and solar energy absorbers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
17194-00-2 | |
171.341 | |
RQPZNWPYLFFXCP-UHFFFAOYSA-L | |
6093286 | |
barium(2+);dihydroxide |
BaH2O2 | |
MFCD00003443 | |
barium hydroxide, barium 2+ dihydroxide, bariumhydroxid, aetzbaryt, barium ii hydroxide, barium 2+ hydroxide, acmc-20alr7, barium hydroxide,anhydrous, barium hydroxide anhydrous | |
CHEBI:32592 | |
[OH-].[OH-].[Ba+2] |
Specifications
408°C | |
Powder | |
BaH2O2 | |
barium hydroxide, barium 2+ dihydroxide, bariumhydroxid, aetzbaryt, barium ii hydroxide, barium 2+ hydroxide, acmc-20alr7, barium hydroxide,anhydrous, barium hydroxide anhydrous | |
RQPZNWPYLFFXCP-UHFFFAOYSA-L | |
barium(2+);dihydroxide | |
6093286 | |
171.36 | |
Barium hydroxide, anhydrous |
17194-00-2 | |
94-98% | |
MFCD00003443 | |
Soluble in water. | |
[OH-].[OH-].[Ba+2] | |
171.341 | |
CHEBI:32592 | |
Air sensitive; Hygroscopic |
Safety and Handling
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H332-H314
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 241-234-5
missing translation for 'rtecsNumber' : CQ9200000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : Air Sensitive & Hygroscopic
Recommended Storage : Ambient temperatures; Store under Argon
- Barium hydroxide, anhydrous
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.