missing translation for 'onlineSavingsMsg'
Learn More

Barium pieces, 2.5cm (1in) & down, 99.2% (metals basis), Sr ∣<0.8%, Thermo Scientific Chemicals

£56.38 - £675.36

Chemical Identifiers

CAS 7440-39-3
Molecular Formula Ba
Molecular Weight (g/mol) 137.33
MDL Number MFCD00134031
InChI Key DSAJWYNOEDNPEQ-UHFFFAOYSA-N
Synonym bario, baryum, bario spanish, baryum french, barium, soluble compounds, unii-24gp945v5t, and soluble compounds, barium, elemental, and compounds, atom
PubChem CID 5355457
ChEBI CHEBI:32595
IUPAC Name barium
SMILES [Ba]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11356607
View Documents
Thermo Scientific Alfa Aesar
000653.06
5 g
£56.38
5g
Please to purchase this item. Need a web account? Register with us today!
11376607
View Documents
Thermo Scientific Alfa Aesar
000653.18
50 g
£151.76
50g
Please to purchase this item. Need a web account? Register with us today!
11366607
View Documents
Thermo Scientific Alfa Aesar
000653.30
250 g
£675.36
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-39-3
137.33
DSAJWYNOEDNPEQ-UHFFFAOYSA-N
5355457
barium
Ba
MFCD00134031
bario, baryum, bario spanish, baryum french, barium, soluble compounds, unii-24gp945v5t, and soluble compounds, barium, elemental, and compounds, atom
CHEBI:32595
[Ba]

Specifications

Silver to Gray
Silver Gray pieces
Ba
bario, baryum, bario spanish, baryum french, barium, soluble compounds, unii-24gp945v5t, and soluble compounds, barium, elemental, and compounds, atom
[Ba]
137.33
CHEBI:32595
Pieces
7440-39-3
99.2% (Metals basis)
MFCD00134031
DSAJWYNOEDNPEQ-UHFFFAOYSA-N
barium
5355457
Odorless
Barium
Videos
Safety and Handling

Safety and Handling

P223-P231+P232-P280-P301+P310-P331-P335+P334-P370+P378q-P501c

H260-H304

missing translation for 'dotInformation' : DOT Class: 4.3, Packaging Group: II

missing translation for 'einecsNumber' : 231-149-1

missing translation for 'rtecsNumber' : CQ8370000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Barium pieces

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 2
  • Substance which\, in contact with water\, emits flammable gases Category 2

Hazard Statement
  • H228-Flammable solid.
  • H261-In contact with water releases flammable gases.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P370+P378-In case of fire: Use to extinguish.
  • P402+P404-Store in a dry place. Store in a closed container.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only