missing translation for 'onlineSavingsMsg'
Learn More
Barium rod, 22mm (0.9in) dia, 99+% (metals basis), Sr ∣<0.8%, Thermo Scientific Chemicals Product Code.: 11351148

Barium rod, 22mm (0.9in) dia, 99+% (metals basis), Sr ∣<0.8%, Thermo Scientific Chemicals

Product Code. 11351148
500 g
Click to view available options
Quantity:
100 g
500 g
1 kg
This item is not returnable. View return policy

Product Code. 11351148

Brand: Thermo Scientific Alfa Aesar 010103.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as getter, in cathode ray tubes, thermionic emitter tubes and sodium vapor lamps

Used as getter, in cathode ray tubes, thermionic emitter tubes and sodium vapor lamps

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-39-3
Molecular Formula Ba
Molecular Weight (g/mol) 137.33
MDL Number MFCD00134031
InChI Key DSAJWYNOEDNPEQ-UHFFFAOYSA-N
Synonym bario, baryum, bario spanish, baryum french, barium, soluble compounds, unii-24gp945v5t, and soluble compounds, barium, elemental, and compounds, atomShow MoreShow Less
PubChem CID 5355457
ChEBI CHEBI:32595
IUPAC Name barium
SMILES [Ba]

Specifications

Color Gray
UN Number UN1400
Quantity 500 g
Percent Purity ≥99%
Concentration or Composition (by Analyte or Components) Sr ≤0.8%
Odor Odorless
Weight ≈7.3cm/100g
Form Rod
Assay (metals basis)
Chemical Name or Material Barium rod, 22mm (0.9 in.) dia.

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR05H113 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog Number010103.22,010103.A1,010103.36
Lot NumberQ05I011 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number010103.22,010103.A1,010103.36
Lot NumberP30H082 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010103.22,010103.A1,010103.36
Lot NumberS17H005 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010103.22,010103.A1,010103.36
Lot NumberD22U008 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010103.22,010103.A1,010103.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Barium rod, 22mm (0.9in) dia, 99+% (metals basis), Sr ∣<0.8%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.