missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Barnstead™ Cartridge Kits

Choose from an array of Thermo Scientific™ Barnstead™ Cartridge Kits for select Thermo Scientific™ Barnstead™ water systems, based on your application.

Brand:  Thermo Scientific™ D3805

535.80 GBP valid until 2025-03-31
Use promo code "24119" to get your promotional price.


 View more versions of this product

Product Code. 10774267

  • £564.00 / Each
Estimated Shipment: 05-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Choose from an array of Thermo Scientific™ Barnstead™ Cartridge Kits for select Thermo Scientific™ Barnstead™ water systems, based on your application.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tap or Pretreated
Nanopure II and older Nanopure system
D3805
Water Purification Cartridge
4.18 cm
8.5 cm
3.8
One 09-034-104 Organic/colloid and two 09-034-3 Ultrapure™ ion exchange cartridges
30 psi
17 in.
3.38 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Up to 60% off
    Promo Code24119

    Expires2025-04-30

    New Offer
    Save Now
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Barnstead™ Cartridge Kits > Nanopure II 3-Module Cartridge Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.