Learn More
Thermo Scientific™ Barnstead™ Cartridge Kits
Choose from an array of Thermo Scientific™ Barnstead™ Cartridge Kits for select Thermo Scientific™ Barnstead™ water systems, based on your application.
£557.60 - £1144.45
Product Code | Brand | For Use With (Equipment) | Feedwater Source | Includes | Model | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Feedwater Source | Includes | Model | Price | Quantity & Availability | ||
10256704
|
Thermo Scientific™
D50254 |
Nanopure Infinity Systems | Deionized, Tap | One 09-050-226 Tap/DI Feed Pretreatment, 09-050-246 Organicfree cartridge, two 09-050-247 Ultrapure™ cartridges and two 09-050-279 Remote Dispenser 0.2μm filters | D50254 |
£1110.03
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
11946092
|
Thermo Scientific™
D5026 |
Nanopure Quad-style system | Pretreated | One 09-034-5C Macropure™ Pretreatment, 09-050-267 Organicfree, 09-050-276 Ultrapure™ and FS090343 Ultrapure™ cartridges | D5026 |
£869.09
Each |
Estimated Shipment: 01-10-2024
Log in to see stock. |
||
11860810
|
Thermo Scientific™
D50255 |
Nanopure Infinity Systems | Reverse Osmosis, Distilled | One 09-050-241 reverse osmosis/Distilled Feed Pretreatment, 09-050-246 Organicfree cartridge, two 09-050-247 Ultrapure™ cartridges and two 09-050-279 Remote Dispenser 0.2μm filters | D50255 |
£1092.82
Each |
In Stock
Log in to see stock. |
||
11756259
|
Thermo Scientific™
D4802 |
Nanopure Quad-style system | Tap | One 09-034-5C Macropure™ Pretreatment, 09-034 high capacity ion exchange, 09-050-276 Ultrapure™ and 09-050-267 Organicfree cartridges | D4802 |
£815.74
Each |
Estimated Shipment: 07-11-2024
Log in to see stock. |
||
10774267
|
Thermo Scientific™
D3805 |
Nanopure II and older Nanopure system | Tap or Pretreated | One 09-034-104 Organic/colloid and two 09-034-3 Ultrapure™ ion exchange cartridges | D3805 |
£557.60
Each |
Estimated Shipment: 07-11-2024
Log in to see stock. |
||
15129583
|
Thermo Scientific™
D3804 |
Nanopure II and older Nanopure system | Tap or Pretreated | One 09-034 high capacity ion exchange, 09-034-104 Organic/colloid and two 09-034-3 Ultrapure™ ion exchange cartridges | D3804 |
£715.07
Each |
Estimated Shipment: 01-10-2024
Log in to see stock. |
||
11840810
|
Thermo Scientific™
D50257 |
Nanopure Infinity Systems | Reverse Osmosis, Distilled | One 09-050-241 reverse osmosis/Distilled Feed Pretreatment, three 09-050-247 Ultrapure™ cartridges, two 09-050-279 remote dispenser 0.2μm filters | D50257 |
£1127.24
Each |
Estimated Shipment: 01-10-2024
Log in to see stock. |
||
11830810
|
Thermo Scientific™
D50256 |
Nanopure Infinity Systems | Deionized, Tap | One 09-050-226 Tap/DI Pretreatment, three 09-050-247 Ultrapure™ cartridges and two 09-050-279 Remote Dispenser 0.2μm filters | D50256 |
£1144.45
Each |
Estimated Shipment: 07-11-2024
Log in to see stock. |
||
11746259
|
Thermo Scientific™
D4801 |
Nanopure Quad-style system | Tap | One 09-034 high capacity ion exchange, 09-034-104 Organic/colloid and two 09-050-276 Ultrapure™ cartridges | D4801 |
£696.99
Each |
Estimated Shipment: 01-10-2024
Log in to see stock. |
||
Description
Choose from an array of Thermo Scientific™ Barnstead™ Cartridge Kits for select Thermo Scientific™ Barnstead™ water systems, based on your application.