Learn More
Thermo Scientific™ Barnstead™ Cartridge Kits
Choose from an array of Thermo Scientific™ Barnstead™ Cartridge Kits for select Thermo Scientific™ Barnstead™ water systems, based on your application.
Brand:  Thermo Scientific™ D50257
 View more versions of this product
Product Code. 11840810
  • £1140.00 / Each
Estimated Shipment: 25-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Choose from an array of Thermo Scientific™ Barnstead™ Cartridge Kits for select Thermo Scientific™ Barnstead™ water systems, based on your application.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Reverse Osmosis, Distilled
Nanopure Infinity Systems
D50257
Water purification cartridge
One 09-050-241 reverse osmosis/Distilled Feed Pretreatment, three 09-050-247 Ultrapure™ cartridges, two 09-050-279 remote dispenser 0.2μm filters
Cartridge Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Barnstead™ Cartridge Kits > Cartridge Kit Type 1 RO/Distilled Water Feed