Learn More
Thermo Scientific™ Barnstead™ Cartridge Kits
Choose from an array of Thermo Scientific™ Barnstead™ Cartridge Kits for select Thermo Scientific™ Barnstead™ water systems, based on your application.
Brand:  Thermo Scientific™ D4802
 View more versions of this product
Product Code. 11756259
  • £824.00 / Each
Estimated Shipment: 05-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Choose from an array of Thermo Scientific™ Barnstead™ Cartridge Kits for select Thermo Scientific™ Barnstead™ water systems, based on your application.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tap
Nanopure Quad-style system
D4802
Water purification cartridge
One 09-034-5C Macropure™ Pretreatment, 09-034 high capacity ion exchange, 09-050-276 Ultrapure™ and 09-050-267 Organicfree cartridges
Cartridge Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Barnstead™ Cartridge Kits > HPLC Grade Cartridge Kit, 4/pk