Learn More
Thermo Scientific™ Barnstead™ Cartridge Kits
Choose from an array of Thermo Scientific™ Barnstead™ Cartridge Kits for select Thermo Scientific™ Barnstead™ water systems, based on your application.
Brand: Thermo Scientific™ D50256
This item is not returnable.
View return policy
Description
Choose from an array of Thermo Scientific™ Barnstead™ Cartridge Kits for select Thermo Scientific™ Barnstead™ water systems, based on your application.
Specifications
Deionized, Tap | |
Nanopure Infinity Systems | |
D50256 |
Water purification cartridge | |
One 09-050-226 Tap/DI Pretreatment, three 09-050-247 Ultrapure™ cartridges and two 09-050-279 Remote Dispenser 0.2μm filters | |
Cartridge Kit |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Thermo Scientific™ Barnstead™ Cartridge Kits > Cartridge Kit Type1 Tap/Distilled Water Feed
Spot an opportunity for improvement?Share a Content Correction