Learn More
MP Biomedicals™ Basic Agarose Premier
Basic Agarose Premier, Analytical Grade
Brand: MP Biomedicals™ 11AGAF0100
87.92 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : CAS Number : 9012-36-6 Weight : 0.10000kg
Description
- Agarose is a purified linear galactan hydrocolloid isolated from agar or agar-bearing marine algae.
- Basic agarose premier is ideal for routine rapid separation of DNA and RNA fragments as well as PCR products, the preparation of plasmids, and for screening, cloning and blotting techniques.
Specifications
9012-36-6 | |
White | |
Powder | |
100 g |
Agarose is a purified lineargalactan hydrocolloid isolated from agar or agar-bearing marine algae. | |
Agarose | |
Analysis of nucleic acids bygel electrophoresis | |
Store at Room Temperature (15-30°C). |