missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Basic Agarose Premier
Basic Agarose Premier, Analytical Grade
£93.49 - £460.54
Specifications
CAS | 9012-36-6 |
---|---|
Description | Agarose is a purified lineargalactan hydrocolloid isolated from agar or agar-bearing marine algae. |
Color | White |
Product Type | Agarose |
Form | Powder |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15792432
|
MP Biomedicals™
11AGAF0100 |
100 g |
£93.49
100g |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
15702442
|
MP Biomedicals™
11AGAF0500 |
500 g |
£460.54
500g |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
Description
- Agarose is a purified linear galactan hydrocolloid isolated from agar or agar-bearing marine algae.
- Basic agarose premier is ideal for routine rapid separation of DNA and RNA fragments as well as PCR products, the preparation of plasmids, and for screening, cloning and blotting techniques.
Specifications
9012-36-6 | |
White | |
Powder | |
Store at Room Temperature (15-30°C). |
Agarose is a purified lineargalactan hydrocolloid isolated from agar or agar-bearing marine algae. | |
Agarose | |
Analysis of nucleic acids bygel electrophoresis |