missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Basic Vortex Mixers

The Thermo Scientific™ Basic Vortex Mixer is ideal for quick, uniform mixing of samples with precise speed control.

£310.22

Specifications

Orbit 4 mm
Load Bearing Capacity (Metric) 0.5 kg
Speed Range 200 to 3000 rpm
Electrical Requirements 100/240 V
Holds Tubes
View More Specs

Includes: Standard package includes the Rubber platform 88882120 x 1

Products 1
Product Code Brand Certifications/Compliance Plug Type Price Quantity & Availability  
Product Code Brand Certifications/Compliance Plug Type Price Quantity & Availability  
17328272
View Documents Promotion Details
Thermo Scientific™
88882012
CE EU, UK, CN Plug
£310.22
Each
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

The Thermo Scientific™ Basic Vortex Mixer provides comprehensive mixing capabilities for enzymatic and RIA assays, general test tube mixing, biochemical assays, viral dilution preparation, precipitated assays, cell suspension vortexing and tissue sample mixing.

  • Digital Speed display 
  • Continuous and Touch modes
  • Speed range: 200 to 3000rpm
  • Speed Accuracy: ± 5% rpm
  • Convenient Speed adjustments by knob control
  • Stable for long-term operations
  • Multiple accessories for different vessels

Order Info

Please order various accessories as the adapter for multiple applications

Specifications

Specifications

4 mm
200 to 3000 rpm
Tubes
ABS housing
11.4 in.
14.99 lb.
14.33 lb.
Basic Vortex Mixer
Accessories: 88882120, 88882121, 88882122, 88882123, 88882124, 88882125, 88882126, 88882127
IP 32
19
2 Years
0.5 kg
100/240 V
50/60 Hz
290 x 270 x 240 mm (11.4 x 10.7 x 9.4 in.)
290 mm
6.8 kg
6.5 kg
Digital
Rubber platform 88882120
DC motor
Touch, Continous
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.