Learn More
Elma Basket System
Modular basket system for Elmasonic P, Elmasonic Select, EASY
Brand:  Elma 1111838
 View more versions of this product
Product Code. 17848835
  • £121.00 / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Stainless steel with plastic handles
  • Ergonomic handles
  • Autoclavable
  • Bayonet holes in the bottom for mounting holders (e.g. Elma Pin, silicone holder bottom comb)
  • Square openings in the side walls for mounting silicone holders
Specifications
Specifications
Modular Basket System
11 lb.
105 x 455 x 55 mm
5.0 kg
Fisherbrand Select. Fisherbrand EASY. Elmasonic EASY. Elmasonic P
Basket System Modular Type 80