missing translation for 'onlineSavingsMsg'
Learn More

Brady™ BBP12 Label printer - UK with Brady Workstation LAB Suite

A compact, reliable, affordable entry level thermal transfer benchtop printer.

Brand:  Brady™ 306869

725.79 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.


 View more versions of this product

Product Code. 17127011

  • £818.00 / 1 set
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The Brady BBP™12 label printer is a compact, reliable, affordable entry level thermal transfer benchtop printer.
  • Ideal for customers requiring low volume printing on lots of different materials within the electrical, voice and datacom, LAB and specialized electronics markets.
  • It allows you to print labels, tapes cable markers, sleeves and much more.
  • The printer comes with the Brady Workstation Laboratory Identification Software suite enabling you to immediately start designing professional labels.
  • This suite combines frequently used software apps to create laboratory identification labels.
Specifications

Specifications

Color LCD
202.00 mm
258.00 mm
Monochrome
Pre-installed cutter; ships with an unwinder, USB Cable, power cord, product CD, Quick Start Guide and Brady Workstation Laboratory ID Suite
300 dpi
Thermal Transfer
173.00 mm
258.00 x 202.00 x 173.00 mm
With Cutter, Unwinder and Brady Workstation LAB Suite
101.60 mm/s
AC Adaptor
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers