Learn More
BD BBL™ Dehydrated Culture Media: Brain Heart Infusion
Used in cultivation of fastidious and nonfastidious microorganisms, including aerobic and anaerobic bacteria,from variety of clinical and nonclinical materials
Brand: BD 211059
Additional Details : CAS Number : 50-99-7 Weight : 0.50000kg
Description
- Formulation: Brain heart infusion from (solids) 6g/L, Peptic digest of Animal Tissue 6g/L, Pancreatic digest of Gelatin 14.5g/L, Dextrose 3g/L, Sodium Chloride 5g/L, Disodium Phosphate 2.5g/L
Certifications
AOAC, BAM, CCAM, CLSI, CMPH2, COMPF, EPA, ISO, MCM9, SMD, SMWW, USDA
Specifications
AOAC, BAM, CCAM, CLSI, CMPH2, COMPF, EPA, ISO, MCM9, SMD, SMWW, USDA | |
Brain Heart Infusion | |
Brain heart infusion from (solids) 6g/L, Peptic digest of Animal Tissue 6g/L, Pancreatic digest of Gelatin 14.5g/L, Dextrose 3g/L, Sodium Chloride 5g/L, Disodium Phosphate 2.5g/L | |
Bottle |
Dehydrated | |
Brain Heart Infusion | |
For fastidious, non-fastidious, aerobic and anaerobic | |
500 g |