missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Prepared Bottled Media: Buffered Sodium Chloride Peptone Solution (pH 7.0)
Used for dissolving, suspending and diluting test samples
Brand: BD 257086
Additional Details : Weight : 3.00000kg
Description
- Used to make suspensions of organisms for testing growth promoting and inhibitory properties of media when examining non-sterile pharmaceutical products for specified microorganisms
- Fluid provides osmotic stability, stable pH value
- Maintains viability of microorganisms during preparation of samples
- Surface-active ingredients or inactivators of antimicrobial agents
Specifications
ISO/DIS, 11014 | |
5°C to 25°C | |
United States | |
Proteose Peptone No.3 10g/L, Potassium Dihydrogen Phosphate 3.6g/L, Disodium Hydrogen Phosphate (Dihydrate) 7.2g/L, Sodium Chloride 4.3g/L | |
Injection stopper and pull ring | |
100 mL |
Clear | |
Liquid | |
Buffered Sodium Chloride Peptone solution | |
Detection of micororganisms from pharmaceutical, food, environmental or Water testing samples | |
Sterile prepared infusion bottle, 25/Pk |