missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Prepared Bottled Media: Buffered Sodium Chloride Peptone Solution (pH 7.0)
Used for dissolving, suspending and diluting test samples
£95.87 - £140.35
Specifications
Color | Clear |
---|---|
Content And Storage | 5°C to 25°C |
Form | Liquid |
Manufacturing Origin | United States |
Media Type | Buffered Sodium Chloride Peptone solution |
Product Code | Brand | Certifications/Compliance | Packaging Type | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Certifications/Compliance | Packaging Type | Quantity | Price | Quantity & Availability | |||
15144172
|
BD
257087 |
USP, EP, JP | Sterile prepared infusion bottle, 10/Pk. | 500 mL |
£95.87
Pack of 10 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||
15124172
|
BD
257086 |
ISO/DIS, 11014 | Sterile prepared infusion bottle, 25/Pk | 100 mL |
£140.35
Pack of 25 |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||
Description
- Used to make suspensions of organisms for testing growth promoting and inhibitory properties of media when examining non-sterile pharmaceutical products for specified microorganisms
- Fluid provides osmotic stability, stable pH value
- Maintains viability of microorganisms during preparation of samples
- Surface-active ingredients or inactivators of antimicrobial agents
Specifications
Clear | |
Liquid | |
Buffered Sodium Chloride Peptone solution | |
Injection stopper and pull ring |
5°C to 25°C | |
United States | |
Detection of micororganisms from pharmaceutical, food, environmental or Water testing samples |