missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Replacement Reagents for Gram Stain Kits
For BBL Gram Stain Kits
Brand: BD 212532
View more versions of this product
This item is not returnable.
View return policy
Description
- Complete stain kits reduce the mess associated with Gram staining procedures
- Break-resistant solution bottle features a hinged spot cap that makes regulating the flow easy and helps prevent spots and spills
- Color-coded labels allow quick identification of reagents
- Stabilized Iodine Solution requires no mixing and has a long shelf life
Order Info
Available in packs of four 250mL bottles or individual one-gallon bottles; sales of B12542 are restricted from California
Specifications
Safranin | |
CE | |
15° to 30°C |
3.8L | |
Used to stain microorganisms from cultures or specimens by the differential gram method | |
Safranin |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Flammable liquid Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P240-Ground/bond container and receiving equipment.
- P233-Keep container tightly closed.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501-Dispose of contents/container to in accordance with all local/regional/national/internationalregulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol, methanol