missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Replacement Reagents for Gram Stain Kits
For BBL Gram Stain Kits
£82.85 - £326.00
Specifications
Certifications/Compliance | CE |
---|---|
For Use With (Application) | Used to stain microorganisms from cultures or specimens by the differential gram method |
Storage Requirements | 15° to 30°C |
Product Code | Brand | Type | Quantity | Stain Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Quantity | Stain Type | Price | Quantity & Availability | |||
11728203
|
BD
212531 |
Safranin | 4 x 250mL | Safranin |
£82.85
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11728213
|
BD
212532 |
Safranin | 3.8L | Safranin |
£281.00
3.80 L |
Estimated Shipment: 13-01-2025
Log in to see stock. |
|||
11758213
|
BD
212542 |
Gram Iodine | 4 x 250mL | Iodine |
£149.00
1000mL |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||
11768213
|
BD
212543 |
Gram Iodine (Stabilized) | 3.8L | Iodine |
£326.00
3.80 L |
Estimated Shipment: 13-01-2025
Log in to see stock. |
|||
11778213
|
BD
212544 |
Basic Fuchsin | 4 x 250mL | Basic Fuchsin |
£174.00
1000mL |
Estimated Shipment: 13-01-2025
Log in to see stock. |
|||
Description
- Complete stain kits reduce the mess associated with Gram staining procedures
- Break-resistant solution bottle features a hinged spot cap that makes regulating the flow easy and helps prevent spots and spills
- Color-coded labels allow quick identification of reagents
- Stabilized Iodine Solution requires no mixing and has a long shelf life
Order Info
Available in packs of four 250mL bottles or individual one-gallon bottles; sales of B12542 are restricted from California
Specifications
CE | |
Used to stain microorganisms from cultures or specimens by the differential gram method | |
15° to 30°C |