missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Replacement Reagents for Gram Stain Kits
For BBL Gram Stain Kits
Brand: BD 212542
This item is not returnable.
View return policy
Description
- Complete stain kits reduce the mess associated with Gram staining procedures
- Break-resistant solution bottle features a hinged spot cap that makes regulating the flow easy and helps prevent spots and spills
- Color-coded labels allow quick identification of reagents
- Stabilized Iodine Solution requires no mixing and has a long shelf life
Order Info
Available in packs of four 250mL bottles or individual one-gallon bottles; sales of B12542 are restricted from California
Specifications
Gram Iodine | |
CE | |
15° to 30°C |
4 x 250mL | |
Used to stain microorganisms from cultures or specimens by the differential gram method | |
Iodine |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P362-Take off contaminated clothing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : polyvinylpyrrolidone-iodine, potassium iodide