Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Erythromycin (15μg)
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
Brand: BD 231290
Additional Details : Weight : 0.80000kg
Description
- Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
Specifications
Erythromycin | |
15μg | |
Disc | |
10/Pk. |