Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Erythromycin (15μg)
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
£46.70
Specifications
Antimicrobial Agents | Erythromycin |
---|---|
Concentration | 15μg |
Packaging Type | Disc |
Description
- Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
Specifications
Erythromycin | |
15μg | |
Disc |