missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Taxo™ P Discs (Vial)
For presumptive identification of pneumococci, and are intended for use with pure cultures
Brand: BD 231047
Additional Details : Weight : 0.80000kg
Description
- 6mm discs made from high quality absorbent paper
- Impregnated with approximately 5.0μg of ethylhydrocupreine hydrochloride per disc
Order Info
Vials of 50 discs.
Specifications
Ethylhydrocupreine hydrochloride (optochin) | |
-20°C to +8°C | |
6 Vials |
5μg | |
Differentiating Pneumococci | |
6 x 50 Discs |
For in vitro Diagnostic Use.