Learn More
BD Biotin Human T Lymphocyte Enrichment Set - DM
For negative selection of T lymphocytes from peripheral blood
Brand: BD 557874
Additional Details : Weight : 0.02000kg
Includes: CD11b/Mac-1, CD16, CD19,CD36, CD41a, CD56, CD235a biotinylated antibody cocktail and BD IMag™ SAv particles
Description
The set components are Biotin Human T Lymphocyte Enrichment Cocktail and Biotin Human T Lymphocyte Enrichment Cocktail
Components
- Biotin Human T Lymphocyte Enrichment Cocktail
- Biotin Human T Lymphocyte Enrichment Cocktail
- Reactivity: Human
Magnetic Labeling
Specifications
Human | |
T-Lymphocyte Enrichment | |
Magnetic Nanoparticles | |
5 mL |
CD11b/Mac-1, CD16, CD19,CD36, CD41a, CD56, CD235a Biotinylated Antibody cocktail and BD IMag™ SAv particles | |
4°C | |
Magnetic labeling |
Research Use Only