Learn More
BD Pharmingen™ Calcein AM Fluorescent Dye
Calcein AM is used for labeling live cells that can be detected and further analyzed by flow cytometry or fluorescence imaging. The enhanced hydrophobicity of the acetomethoxy (AM) derivative of Calcein allows this dye to readily enter viable cells.
Brand: BD 564061
149.71 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The enhanced hydrophobicity of the acetomethoxy (AM) derivative of Calcein allows this dye to readily enter viable cells. Once inside, intracellular esterases cleave the AM groups off of the non-fluorescent BD Pharmingen™ Calcein AM, trapping fluorescent Calcein within the cell. Since dead cells lack esterase activity, only viable cells are labeled. Calcein AM is optimally excited at the 495 nm wavelength of light and emits maximally at 515 nm.
Specifications
Calcein AM fluorescent dye | |
Fluorescent Dye | |
RUO |
Flow Cytometry, Bioimaging | |
1 mg |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.