missing translation for 'onlineSavingsMsg'
Learn More

BD Cell Separation Magnet

Specialized plastic test-tube rack surrounding a strong permanent rare earth magnet

Brand:  BD 552311

Additional Details : Weight : 1.10000kg

Product Code. 15971260

  • £882.00 / Each
Estimated Shipment: 26-07-2024
to see stock.
Add to basket

Alert:

The BD IMagnet™ contains a strong permanent magnet that may affect or damage delicate instruments, electronic equipment, and magnetic recording devices. Therefore, persons wearing cardiac pacemakers should not handle the BD IMagnet, and the BD IMagnet should not be placed near laboratory instruments, monitors, computer discs, credit cards, video or audio cassettes, and watches. The BD IMagnet has a very strong attraction for ferric metals, so it should not be placed on steel surfaces or near objects containing steel or iron.


Description

Description

  • Holds up to six 12 x 75mm or two 17 x 100mm round-bottom test tubes
  • Allows rapid and effective positive or negative selection of desired leukocyte populations

Cell separation

Specifications

Specifications

Each
Cell separation
Cell Separation Magnet
Six 12 x 75mm/two 17 x 100mm round-botto m test tubes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For research use only.