missing translation for 'onlineSavingsMsg'
Learn More

BD Cell Signaling Master Buffer Kit

Employ particles with discrete fluorescence intensities to detect soluble analytes at very low concentrations

Brand:  BD 560006

Additional Details : Weight : 1.90100kg

 View more versions of this product

Product Code. 15809868

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Includes:
  • Assay diluent
  • Capture bead diluent
  • Detection reagent diluent
  • Denaturation buffer
  • Wash buffer
  • Instrument setup reagents

Description

Description

The buffers and instrument setup reagents provided in this kit have been optimized to measure total or phosphorylated proteins from denatured cell lysate sample.

  • Contains all of the supporting reagents necessary to perform an assay using a BD CBA Cell Signaling Flex Set
  • Provides sufficient reagents for the quantitative analysis of 100 samples and 10 instrument setup procedures
  • Working assay range for most analytes in this system is 3.9 to 1,000 units/mL

For research use only. Not for use in diagnostic or therapeutic procedures.

Specifications

Specifications

1 bottle 150mL assay diluent, 1 bottle 30mL capture bead diluent, 1 bottle 30mL detection reagent diluent, 1 bottle 150mL denatuRation buffer 5X, 1 bottle 650mL wash buffer, 1 vial 0.25mL instrument setup bead A1, 1 vial 0.25mL instrument setup bead A9
500
RUO
2°C to 8°C
Cytometric Bead Array (CBA) Assay
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers