missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cell Signaling Master Buffer Kit
Employ particles with discrete fluorescence intensities to detect soluble analytes at very low concentrations
£463.62 - £1765.00
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Product Type | Cytometric Bead Array (CBA) Assay |
Validated Application | RUO |
Includes:
- Assay diluent
- Capture bead diluent
- Detection reagent diluent
- Denaturation buffer
- Wash buffer
- Instrument setup reagents
Product Code | Brand | Includes | No. of Tests | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | No. of Tests | Price | Quantity & Availability | ||||
15899858
|
BD
560005 |
1 bottle 30mL assay diluent, 1 bottle 5mL capture bead diluent, 1 bottle 5mL detection reagent diluent, 1 bottle 30mL denaturation buffer 5X, 1 bottle 130mL wash buffer, 1 vial 0.25mL instrument setup bead A1, 1 vial 0.25mL instrument setup bead A9 | 100 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15809868
|
BD
560006 |
1 bottle 150mL assay diluent, 1 bottle 30mL capture bead diluent, 1 bottle 30mL detection reagent diluent, 1 bottle 150mL denatuRation buffer 5X, 1 bottle 650mL wash buffer, 1 vial 0.25mL instrument setup bead A1, 1 vial 0.25mL instrument setup bead A9 | 500 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
The buffers and instrument setup reagents provided in this kit have been optimized to measure total or phosphorylated proteins from denatured cell lysate sample.
- Contains all of the supporting reagents necessary to perform an assay using a BD CBA Cell Signaling Flex Set
- Provides sufficient reagents for the quantitative analysis of 100 samples and 10 instrument setup procedures
- Working assay range for most analytes in this system is 3.9 to 1,000 units/mL
For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications
2°C to 8°C | |
Cytometric Bead Array (CBA) Assay | |
RUO |